Invitrogen
Manufacturer Code:PA552847
Catalog # PIPA552847
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SELENBP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 56 a selenium binding protein hSBP Lp 56 Lp56 Lpsb SBP 1 SBP SBP1 SBP56 Selen bp2 SELENBP 1 Selenbp 2 Selenbp2 Selenium binding protein 2 Selenium binding protein1 Selenium binding protein2 SELNBP1 SP 56 SP56; 56 kDa selenium-binding protein; epididymis secretory sperm binding protein Li 134P; Methanethiol oxidase; MTO; SBP56; Selenium-binding protein 1
Gene Aliases: HEL-S-134P; hSBP; LPSB; SBP; SBP56; SELENBP1; SP56
UniProt ID: (Human) Q13228
Entrez Gene ID: (Human) 8991
Molecular Function: defense/immunity protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.