Invitrogen
Manufacturer Code:PA555035
Catalog # PIPA555035
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SEL1L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein sel-1 homolog 1; sel-1 suppressor of lin-12-like 1; Sel-1L; SEL1L sel-1 suppressor of lin-12-like (C. elegans) IBD2 PRO1063 SEL1-LIKE Suppressor of lin 12 (sel-1) C. elegans homolog of sel-1 (suppressor of lin-12 C.elegans)-like sel-1 suppressor of lin-12-like; Suppressor of lin 12 (sel-1), C. elegans, homolog of; Suppressor of lin-12-like protein 1
Gene Aliases: PRO1063; SEL1-LIKE; SEL1L; SEL1L1; TSA305; UNQ128/PRO1063
UniProt ID: (Human) Q9UBV2
Entrez Gene ID: (Human) 6400
Molecular Function: enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.