Invitrogen
Manufacturer Code:PA5109312
Catalog # PIPA5109312
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein within zebrafish SEL1L aa 100-300. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AW493766 fj60e07 IBD2 mKIAA4137 PRO1063 Protein sel-1 homolog 1 Sel1h SEL1L Sel-1L SEL1L1 SEL1-LIKE si:dkey-177p2.3 Suppressor of lin-12-like protein 1 TSA305 UNQ128/PRO1063 wu:fj60e07 zgc:56075; sel-1 suppressor of lin-12-like; SEL1L, ERAD E3 ligase adaptor subunit
Gene Aliases: fj60e07; si:dkey-177p2.3; wu:fj60e07; zgc:56075
Entrez Gene ID: (Zebrafish) 568615
Molecular Function: enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.