Invitrogen
Manufacturer Code:PA559012
Catalog # PIPA559012
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SEH1L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GATOR complex protein SEH1; Nucleoporin SEH1; Nup107-160 subcomplex subunit SEH1; sec13 like protein; SEC13-like protein; SEH1-like nucleoporin
Gene Aliases: SEC13L; SEH1; SEH1A; SEH1B; SEH1L
UniProt ID: (Human) Q96EE3
Entrez Gene ID: (Human) 81929
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.