Novus Biologicals
Manufacturer Code:NBP222301
Catalog # NBP222301
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen this antibody was made to maps to a region between residue 804 to 854 of human SECIS Binding Protein 2 using the numbering given in entry NP_076982.3 (GeneID 79048). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686C09169 SBP2SECIS-binding protein 2 SECIS binding protein 2 selenocysteine insertion sequence binding protein 2 selenocysteine insertion sequence-binding protein 2; SECIS-binding protein 2; Selenocysteine insertion sequence-binding protein 2
Gene Aliases: SBP2; SECISBP2
UniProt ID: (Human) Q7L1Z0
Entrez Gene ID: (Human) 79048
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.