Invitrogen
Manufacturer Code:PA542936
Catalog # PIPA542936
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human SEC63 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SEC63 homolog; SEC63 homolog (S. cerevisiae) ERdj2 PRO2507 SEC63L DNAJC23; SEC63 protein translocation regulator; SEC63-like protein; Translocation protein SEC63 homolog
Gene Aliases: 5730478J10Rik; AI649014; AW319215; DNAJC23; ERdj2; PRO2507; SEC63; SEC63-like; SEC63L; zgc:92718
UniProt ID: (Human) Q9UGP8, (Mouse) Q8VHE0
Entrez Gene ID: (Dog) 475016, (Horse) 100072096, (Human) 11231, (Bovine) 541040, (Rabbit) 100328696, (Rat) 309858, (Mouse) 140740, (Zebrafish) 436861
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.