Novus Biologicals
Manufacturer Code:NBP286797
Catalog # NB034419
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human SEC61A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSEC61 protein transport protein SEC61 alpha subunit protein transport protein Sec61 subunit alpha protein transport protein Sec61 subunit alpha isoform 1 SEC61 Sec61 alpha 1 subunit (S. cerevisiae) Sec61 alpha-1 sec61 homolog SEC61A; protein transport protein SEC61 alpha subunit; protein transport protein Sec61 subunit alpha; Protein transport protein Sec61 subunit alpha isoform 1; Sec61 alpha 1 subunit; Sec61 alpha-1; sec61 homolog
Gene Aliases: HSEC61; SEC61; SEC61A; SEC61A1
UniProt ID: (Human) P61619
Entrez Gene ID: (Human) 29927
Molecular Function: membrane traffic protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.