Proteintech
Manufacturer Code:147031AP150UL
Catalog # 50-172-9655
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SEC6 Fusion Protein Ag6383 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EXOC3 exocyst complex component 3 Exocyst complex component Sec6 SEC6 SEC6L1 Sec6p; Exocyst complex component 3; Exocyst complex component Sec6; rSec6; Sec 6 homolog; SEC6-like 1
Gene Aliases: 2810050O03Rik; E430013E20Rik; EXOC3; rSec6; SEC6; SEC6L1; Sec6p
UniProt ID: (Human) O60645, (Rat) Q62825, (Mouse) Q6KAR6
Entrez Gene ID: (Human) 11336, (Rat) 252881, (Mouse) 211446
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.