Proteintech
Manufacturer Code:127511AP150UL
Catalog # 50-172-8221
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SEC5/EXOC2 Fusion Protein Ag3147 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EXOC2 exocyst complex component 2 Exocyst complex component Sec5 SEC5 SEC5L1 Sec5p; Exocyst complex component 2; Exocyst complex component Sec5; rSec5; SEC5 homolog; SEC5-like 1
Gene Aliases: 2410030I24Rik; AI648199; EXOC2; Rsec5; SEC5; SEC5L1; Sec5p
UniProt ID: (Human) Q96KP1, (Mouse) Q9D4H1, (Rat) O54921
Entrez Gene ID: (Human) 55770, (Mouse) 66482, (Rat) 171455
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.