Novus Biologicals
Manufacturer Code:NBP29481420UL
Catalog # NB024139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 645-894 of human EXOC1 (NP_001020095.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BM-102 exocyst complex component 1 Exocyst complex component Sec3 FLJ10893 SEC3L1 SEC3-like 1 SEC3-like 1 (S. cerevisiae) SEC3P SEC3Sec3p; Exocyst complex component 1; Exocyst complex component Sec3; SEC3-like 1
Gene Aliases: BM-012; BM-102; EXOC1; SEC3; SEC3L1; SEC3P
UniProt ID: (Human) Q9NV70
Entrez Gene ID: (Human) 55763
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.