Novus Biologicals
Manufacturer Code:NBP186263
Catalog # NBP186263
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SFQGAASSASHLHTSASQPYSSFVNHYNSPAMYSASSSVASQGFPSTCGHYAMSTVSNAAYPSVSYPSLPAGDTYGQMFTSQNAPTVR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: member B MGC48822 protein transport protein Sec24B SEC24 SEC24 family member B (S. cerevisiae) Sec24-related protein B secretory protein 24; Protein transport protein Sec24B; SEC24 related gene family, member B; SEC24-related protein B; secretory protein 24
Gene Aliases: SEC24; SEC24B
UniProt ID: (Human) O95487
Entrez Gene ID: (Human) 10427
Molecular Function:
membrane traffic protein
vesicle coat protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.