Proteintech
Manufacturer Code:208921AP150UL
Catalog # 50-173-3141
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SEC23IP Fusion Protein Ag14916 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MSTP053 P125 SEC23 interacting protein SEC23IP; p125; SEC23-interacting protein
Gene Aliases: D7Ertd373e; MSTP053; P125; P125A; SEC23IP
UniProt ID: (Human) Q9Y6Y8
Entrez Gene ID: (Human) 11196, (Mouse) 207352, (Rat) 309010
Molecular Function:
hydrolase
lipase
membrane traffic protein
phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.