Novus Biologicals
Manufacturer Code:NB10091277
Catalog # NB10091277
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide as a part of human rat mouse hamster orangutan chicken Vesicle-trafficking protein SEC22b (SEC22B SEC22L1 ERS24) conjugated to an immunogenic carrier protein was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ER-Golgi SNARE of 24 kDa; ERS-24; ERS24 ERS-24sec22b SEC22 vesicle trafficking protein homolog B (S. cerevisiae) SEC22 vesicle trafficking protein homolog B (S. cerevisiae) (gene/pseudogene) SEC22 vesicle trafficking protein-like 1 SEC22 vesicle trafficking protein-like 1 (S. cerevisiae) SEC22 vesicle-trafficking protein homolog B SEC22 vesicle-trafficking protein-like 1 SEC22 vesicle trafficking protein (S. cerevisiae)-like 1 SEC22L1ER-Golgi SNARE of 24 kDa vesicle-trafficking protein SEC22b; SEC22 vesicle trafficking protein homolog B; SEC22 vesicle trafficking protein-like 1; SEC22 vesicle-trafficking protein homolog B; SEC22 vesicle-trafficking protein-like 1; Vesicle-trafficking protein SEC22b
Gene Aliases: ERS-24; SEC22B; SEC22L1
UniProt ID: (Human) O75396
Entrez Gene ID: (Human) 9554
Molecular Function: SNARE protein membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.