Proteintech
Manufacturer Code:147761AP150UL
Catalog # 50-172-9711
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SEC22B Fusion Protein Ag6077 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ER Golgi SNARE of 24 kDa ERS 24 ERS24 SEC22B SEC22L1; ER-Golgi SNARE of 24 kDa; ERS-24; ERS24; mSec22b; SEC22 vesicle trafficking protein homolog B; SEC22 vesicle trafficking protein-like 1; SEC22 vesicle-trafficking protein homolog B; SEC22 vesicle-trafficking protein-like 1; Vesicle-trafficking protein SEC22b
Gene Aliases: 4930564D15Rik; AA517334; AI480645; C81333; ERS-24; SEC22B; SEC22L1
UniProt ID: (Human) O75396, (Rat) Q4KM74, (Mouse) O08547
Entrez Gene ID: (Human) 9554, (Rat) 310710, (Mouse) 20333
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.