Abnova
Manufacturer Code:PAB19201
Catalog # 89-120-731
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 18 amino acids near C-terminus of human SEC16B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody rasied against synthetic peptide of SEC16B.
For Research Use Only
Protein Aliases: DKFZp686C2486; FLJ23871; FLJ25761; FLJ33652; FLJ36620; Leucine zipper transcription regulator 2; LZTR2; PGPR-p117; protein SEC16 homolog B; Protein transport protein Sec16B; Regucalcin gene promoter region-related protein p117; regucalcin gene promotor region related protein; RGPR; RGPR-p117; SEC16 homolog B; SEC16S
Gene Aliases: KIAA1928; LZTR2; PGPR-p117; RGPR; SEC16B; SEC16S
UniProt ID: (Human) Q96JE7
Entrez Gene ID: (Human) 89866
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.