OriGene
Manufacturer Code:TA503561
Catalog # 50-167-4422
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1H3 |
Immunogen | Full length human recombit protein of human SEC14L2 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Alpha-tocopherol-associated protein; C22orf6 SPF TAP TAP1; SEC14-like lipid binding 2; SEC14-like protein 2; SPF; Squalene transfer protein; Supernatant protein factor; TAP; tocopherol-associated protein; tocopherol-associated protein 1
Gene Aliases: C22orf6; KIAA1186; KIAA1658; SEC14L2; SPF; TAP; TAP1
UniProt ID: (Human) O76054
Entrez Gene ID: (Human) 23541
Molecular Function: transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.