Invitrogen
Manufacturer Code:PA576607
Catalog # PIPA576607
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human SEC14L2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-tocopherol-associated protein; SEC14-like lipid binding 2; SEC14-like protein 2; SPF; SPF TAP TAP1 C22orf6; Squalene transfer protein; Supernatant protein factor; TAP; tocopherol-associated protein; tocopherol-associated protein 1
Gene Aliases: 1300013M05Rik; C22orf6; KIAA1186; KIAA1658; SEC14L2; SPF; TAP; TAP1
UniProt ID: (Human) O76054, (Mouse) Q99J08
Entrez Gene ID: (Human) 23541, (Mouse) 67815, (Rat) 116486
Molecular Function: transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.