Novus Biologicals
Manufacturer Code:NBP239029
Catalog # NBP239029
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: ELLLQECAHQTTKLGRKVETITIIYDC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-tocopherol-associated protein; C22orf6 hTAP KIAA1186SEC14 (S. cerevisiae)-like 2 KIAA1658SPFAlpha-tocopherol-associated protein SEC14-like 2 (S. cerevisiae) SEC14-like protein 2 Squalene transfer protein Supernatant protein factor TAP1 TAPMGC65053 tocopherol-associated protein; SEC14-like lipid binding 2; SEC14-like protein 2; SPF; Squalene transfer protein; Supernatant protein factor; TAP; tocopherol-associated protein; tocopherol-associated protein 1
Gene Aliases: C22orf6; KIAA1186; KIAA1658; SEC14L2; SPF; TAP; TAP1
UniProt ID: (Human) O76054
Entrez Gene ID: (Human) 23541
Molecular Function: transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.