Novus Biologicals
Manufacturer Code:MAB90551AF594
Catalog # MB90551A594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1280B |
Immunogen | rhSEC13 aa123-322 |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
Protein Aliases: D3S1231ESEC13L1npp-20 protein SEC13 homolog SEC13 (S. cerevisiae)-like 1 SEC13 homolog (S. cerevisiae) SEC13L1 SEC13-like 1 isoform SEC13-like protein 1 SEC13-related protein SEC13RSEC13-like 1 (S. cerevisiae); GATOR complex protein SEC13; Protein SEC13 homolog; SEC13 homolog; SEC13 homolog, nuclear pore and COPII coating complex component; SEC13-like 1 isoform; SEC13-like protein 1; SEC13-related protein
Gene Aliases: D3S1231E; npp-20; SEC13; SEC13A; SEC13L1; SEC13R
UniProt ID: (Human) P55735
Entrez Gene ID: (Human) 6396
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.