Invitrogen
Manufacturer Code:MA526076
Catalog # PIMA526076
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1G4 |
Immunogen | Full length human recombit protein of SDSL produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: L-serine deaminase; L-serine dehydratase/L-threonine deaminase; L-threonine dehydratase; SDH 2; SDSL SDH 2 SDS-RS1 TDH cSDH; Serine dehydratase 2; serine dehydratase related sequence 1; Serine dehydratase-like; TDH
Gene Aliases: cSDH; SDH 2; SDS-RS1; SDSL; TDH
UniProt ID: (Human) Q96GA7
Entrez Gene ID: (Human) 113675
Molecular Function:
deaminase
dehydratase
epimerase/racemase
hydrolase
isomerase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.