OriGene
Manufacturer Code:TA502015
Catalog # 50-167-3103
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI10F11 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-313 of human SDR9C7 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ARCI13 RDHS SDR-O SDRO; orphan short-chain dehydrogenase / reductase; Orphan short-chain dehydrogenase/reductase; RDH-S; retinol dehydrogenase similar protein; SDR-O; Short-chain dehydrogenase/reductase family 9C member 7
Gene Aliases: RDHS; SDR-O; SDR9C7; SDRO
UniProt ID: (Human) Q8NEX9
Entrez Gene ID: (Human) 121214
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.