Abnova
Manufacturer Code:H00195814K
Catalog # 89-204-632
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SDR16C5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SDR16C5 peptide using ARM Technology.
For Research Use Only
Protein Aliases: EPHD-2; epidermal retinal dehydrogenase 2; Epidermal retinol dehydrogenase 2; FLJ33105 RDH&num2 RDH-E2 RDHE2; retinal short chain dehydrogenase reductase; Retinal short-chain dehydrogenase reductase 2; retSDR2; Short-chain dehydrogenase/reductase family 16C member 5
Gene Aliases: EPHD-2; RDH#2; RDH-E2; RDHE2; retSDR2; SDR16C5
UniProt ID: (Human) Q8N3Y7
Entrez Gene ID: (Human) 195814
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.