Invitrogen
Manufacturer Code:PA534387
Catalog # PIPA534387
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 15 amino acid synthetic peptide near the center of human SDHD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CII-4; CybS; QPs3; SDHD Succinate dehydrogenase complex subunit D succinate dehydrogenase cytochrome b small subunit mitochondrial SDH4 PGL PGL1 CBT1; Succinate dehydrogenase [ubiquinone] cytochrome b small subunit, mitochondrial; Succinate dehydrogenase complex subunit D; succinate dehydrogenase complex subunit D integral membrane protein; Succinate-ubiquinone oxidoreductase cytochrome b small subunit; Succinate-ubiquinone reductase membrane anchor subunit
Gene Aliases: 3110001M13Rik; AVLL5809; C78570; CBT1; CII-4; CWS3; cybS; PGL; PGL1; PRO19626; QPs3; SDH4; SDHD
UniProt ID: (Human) O14521, (Rat) Q6PCT8, (Mouse) Q9CXV1
Entrez Gene ID: (Human) 6392, (Rat) 363061, (Mouse) 66925
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.