Novus Biologicals
Manufacturer Code:NBP281948
Catalog # NB017259
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SDHD antibody was raised against a 15 amino acid synthetic peptide near the center of human SDHD. The immunogen is located within amino acids 30 - 80 of SDHD. Amino Acid Squence: HLSPSHHSGSKAASL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CBT1 CII-4 CybS PGL PGL1 QPs3 SDH4 succinate dehydrogenase [ubiquinone] cytochrome b small subunit mitochondrial Succinate dehydrogenase complex subunit D succinate dehydrogenase complex subunit D integral membrane protein succinate dehydrogenase ubiquinone cytochrome B small subunit Succinate-ubiquinone oxidoreductase cytochrome b small subunit Succinate-ubiquinone reductase membrane anchor subunit; CII-4; CybS; QPs3; Succinate dehydrogenase [ubiquinone] cytochrome b small subunit, mitochondrial; Succinate dehydrogenase complex subunit D; succinate dehydrogenase complex subunit D integral membrane protein; Succinate-ubiquinone oxidoreductase cytochrome b small subunit; Succinate-ubiquinone reductase membrane anchor subunit
Gene Aliases: CBT1; CII-4; CWS3; cybS; PGL; PGL1; QPs3; SDH4; SDHD
UniProt ID: (Human) O14521
Entrez Gene ID: (Human) 6392
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.