Abnova
Manufacturer Code:H00006392K
Catalog # 89-202-504
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SDHD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SDHD peptide using ARM Technology.
For Research Use Only
Protein Aliases: CBT1 PGL PGL1 SDH4; CII-4; CybS; QPs3; Succinate dehydrogenase [ubiquinone] cytochrome b small subunit, mitochondrial; Succinate dehydrogenase complex subunit D; succinate dehydrogenase complex subunit D integral membrane protein; Succinate-ubiquinone oxidoreductase cytochrome b small subunit; Succinate-ubiquinone reductase membrane anchor subunit
Gene Aliases: CBT1; CII-4; CWS3; cybS; PGL; PGL1; QPs3; SDH4; SDHD
UniProt ID: (Human) O14521
Entrez Gene ID: (Human) 6392
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.