Novus Biologicals
Manufacturer Code:NBP286795
Catalog # NB034417
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of SDHC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CYBL; cytochrome B large subunit of complex II; Integral membrane protein CII-3; Integral membrane protein CII-3 mitochondrial PGL3 QPs1 QPs-1 SDH3 succinate dehydrogenase complex subunit C integral membrane protein 15kD succinate dehydrogenase complex subunit C integral membrane protein 15kDa succinate-ubiquinone oxidoreducatase cytochrome B large subunit; integral membrane protein CII-3b; large subunit of cytochrome b; QPs-1; QPs1; succinate dehydrgenase cytochrome b; succinate dehydrogenase 3, integral membrane subunit; Succinate dehydrogenase complex subunit C; succinate dehydrogenase complex subunit C integral membrane protein 15kDa; succinate dehydrogenase complex, subunit C, integral membrane protein, 15kD; succinate dehydrogenase complex, subunit C, integral membrane protein, 15kDa; Succinate dehydrogenase cytochrome b560 subunit, mitochondrial; Succinate-ubiquinone oxidoreductase cytochrome B large subunit
Gene Aliases: CYB560; CYBL; PGL3; QPS1; SDH3; SDHC
UniProt ID: (Human) Q99643
Entrez Gene ID: (Human) 6391
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.