Abnova
Manufacturer Code:PAB25763
Catalog # 89-127-387
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 16 amino acids at N-terminus of human SDHAF2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Chicken polyclonal antibody raised against synthetic peptide of SDHAF2.
For Research Use Only
Protein Aliases: C11orf79; hSDH5; PGL2; SDH assembly factor 2; SDH5; SDHAF2; Succinate dehydrogenase assembly factor 2, mitochondrial; succinate dehydrogenase subunit 5, mitochondrial
Gene Aliases: C11orf79; PGL2; SDH5; SDHAF2
UniProt ID: (Human) Q9NX18
Entrez Gene ID: (Human) 54949
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.