Invitrogen
Manufacturer Code:PA579962
Catalog # PIPA579962
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human SDHA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Flavoprotein subunit of complex II; Fp; Succinate dehydrogenase &lsqbubiquinone] flavoprotein subunit mitochondrial; succinate dehydrogenase [ubiquinone] flavoprotein subunit; Succinate dehydrogenase [ubiquinone] flavoprotein subunit, mitochondrial; succinate dehydrogenase complex subunit A, flavoprotein (Fp); succinate dehydrogenase complex, subunit A, flavoprotein (Fp)
Gene Aliases: 1500032O14Rik; 2310034D06Rik; 4921513A11; C81073; CMD1GG; FP; PGL5; SDH1; SDH2; SDHA; SDHF
UniProt ID: (Human) P31040, (Rat) Q920L2, (Mouse) Q8K2B3
Entrez Gene ID: (Human) 6389, (Rat) 157074, (Mouse) 66945
Molecular Function: dehydrogenase oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.