Novus Biologicals
Manufacturer Code:NBP29392220UL
Catalog # NB029703
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 385-664 of human SDHA (NP_004159.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CMD1GG EC 1.3.5.1 Fp SDH1 SDH2 SDHFmitochondrial succinate dehydrogenase complex flavoprotein subunit succinate dehydrogenase complex subunit A flavoprotein (Fp); Flavoprotein subunit of complex II; Fp; succinate dehydrogenase [ubiquinone] flavoprotein subunit; Succinate dehydrogenase [ubiquinone] flavoprotein subunit, mitochondrial; succinate dehydrogenase complex subunit A, flavoprotein (Fp); succinate dehydrogenase complex, subunit A, flavoprotein (Fp)
Gene Aliases: CMD1GG; FP; PGL5; SDH1; SDH2; SDHA; SDHF
UniProt ID: (Human) P31040
Entrez Gene ID: (Human) 6389
Molecular Function: dehydrogenase oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.