Invitrogen
Manufacturer Code:PA543212
Catalog # PIPA543212
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SDF2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SDF-2; Stromal cell-derived factor 2; stromal cell-derived factor 2-like protein
Gene Aliases: AI853825; fk23f01; SDF2; wu:fk23f01; zgc:66291
UniProt ID: (Bovine) Q3SZ45, (Human) Q9BQ79, (Mouse) Q9DCT5
Entrez Gene ID: (Rabbit) 100345876, (Goat) 102189240, (Bovine) 508463, (Human) 6388, (Dog) 480626, (Horse) 100059307, (Mouse) 20316, (Rat) 287470, (Guinea pig) 100724752, (Zebrafish) 336879
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.