Novus Biologicals
Manufacturer Code:NBP15669920UL
Catalog # NBP15669920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SDCCAG8(serologically defined colon cancer antigen 8) The peptide sequence was selected from the middle region of SDCCAG8. Peptide sequence IQCDQLRKELERQAERLEKELASQQEKRAIEKDMMKKEITKEREYMGSKM. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antigen NY-CO-8; CCCAPAntigen NY-CO-8 Centrosomal colon cancer autoantigen protein hCCCAP NPHP10 NY-CO-8 serologically defined colon cancer antigen 8 SLSN7; Centrosomal colon cancer autoantigen protein; hCCCAP; Serologically defined colon cancer antigen 8
Gene Aliases: BBS16; CCCAP; CCCAP SLSN7; hCCCAP; HSPC085; NPHP10; NY-CO-8; SDCCAG8; SLSN7
UniProt ID: (Human) Q86SQ7
Entrez Gene ID: (Human) 10806
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.