Proteintech
Manufacturer Code:134711AP150UL
Catalog # 50-172-8769
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SDCCAG8 Fusion Protein Ag4264 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antigen NY CO 8 BBS16 CCCAP hCCCAP HSPC085 NY CO 8 SDCCAG8; Antigen NY-CO-8; Centrosomal colon cancer autoantigen protein; hCCCAP; mCCCAP; Serologically defined colon cancer antigen 8; Serologically defined colon cancer antigen 8 homolog; slinky
Gene Aliases: 2700048G21Rik; 5730470G24Rik; BBS16; CCCAP; CCCAP SLSN7; hCCCAP; HSPC085; NPHP10; NY-CO-8; SDCCAG8; si:dkey-60b15.1; SLSN7
UniProt ID: (Human) Q86SQ7, (Mouse) Q80UF4
Entrez Gene ID: (Human) 10806, (Dog) 490374, (Rat) 305002, (Mouse) 76816, (Zebrafish) 556980
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.