Abnova
Manufacturer Code:PAB20340
Catalog # 89-121-840
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human SDC1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant SDC1.
For Research Use Only
Protein Aliases: CD138; CD138 antigen; heparan sulfate proteoglycan fibroblast growth factor receptor; SDC; SYND1; syndecan; syndecan proteoglycan 1; Syndecan-1
Gene Aliases: CD138; SDC; SDC1; SYND1; syndecan
UniProt ID: (Human) P18827
Entrez Gene ID: (Human) 6382
Molecular Function:
cell adhesion molecule
cytoskeletal protein
defense/immunity protein
extracellular matrix glycoprotein
extracellular matrix protein
membrane-bound signaling molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.