Invitrogen
Manufacturer Code:PA589052
Catalog # PIPA589052
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-300 of human SCYL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Coated vesicle-associated kinase of 104 kDa; SCY1-like protein 2
Gene Aliases: AU015688; AW536445; BC030932; CVAK104; D10Ertd802e; RGD1305665; Scyl2
UniProt ID: (Mouse) Q8CFE4
Entrez Gene ID: (Rat) 314717, (Mouse) 213326
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.