Invitrogen
Manufacturer Code:PA5116213
Catalog # PIPA5116213
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 585-613 amino acids from the Central region of mouse Scyl2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AU015688 AW536445 BC030932 Coated vesicle-associated kinase of 104 kDa Cvak104 D10Ertd802e KIAA1360 RGD1305665 SCY1 like pseudokinase 2 SCY1-like 2 SCY1-like 2 (S. cerevisiae) SCY1-like protein 2 SCY1-like kinase-like 2 SCYL2; Coated vesicle-associated kinase of 104 kDa; SCY1-like 2; SCY1-like protein 2; SCY1-like, kinase-like 2
Gene Aliases: AU015688; AW536445; BC030932; CVAK104; D10Ertd802e; KIAA1360; SCYL2
UniProt ID: (Human) Q6P3W7, (Mouse) Q8CFE4
Entrez Gene ID: (Human) 55681, (Mouse) 213326
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.