Novus Biologicals
Manufacturer Code:NBP29362220UL
Catalog # NB029109
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human SCYL2 (NP_060458.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Coated vesicle-associated kinase of 104 kDa; CVAK104 FLJ10074 KIAA1360Coated vesicle-associated kinase of 104 kDa SCY1-like 2 (S. cerevisiae) SCY1-like protein 2; SCY1-like 2; SCY1-like protein 2; SCY1-like, kinase-like 2
Gene Aliases: CVAK104; KIAA1360; SCYL2
UniProt ID: (Human) Q6P3W7
Entrez Gene ID: (Human) 55681
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.