Invitrogen
Manufacturer Code:PA567935
Catalog # PIPA567935
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human SCRIB. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: circletail; CRIB1 SCRB1 SCRIB1 Vartul protein scribble homolog; PDZ-domain protein scribble; Protein LAP4; Protein scribble homolog; Scribble; scribble homolog 1; scribbled homolog
Gene Aliases: AI118201; Crc; CRIB; CRIB1; KIAA0147; LAP4; mKIAA0147; RGD1565055; SCRB1; SCRIB; SCRIB1; VARTUL
UniProt ID: (Human) Q14160, (Mouse) Q80U72
Entrez Gene ID: (Human) 23513, (Mouse) 105782, (Rat) 362938
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.