Abnova
Manufacturer Code:PAB15708
Catalog # 89-117-773
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant GST fusion protein corresponding to 143 mouse Scrib. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant Scrib.
For Research Use Only
Protein Aliases: AI118201; circletail; Crc; CRIB; KIAA0147; mKIAA0147; PDZ-domain protein scribble; Protein LAP4; Protein scribble homolog; Scrb1; SCRIB1; Scribble; scribble homolog 1; vartul
Gene Aliases: AI118201; Crc; CRIB; Kiaa0147; Lap4; mKIAA0147; Scrb1; Scrib; SCRIB1; vartul
UniProt ID: (Mouse) Q80U72
Entrez Gene ID: (Mouse) 105782
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.