Novus Biologicals
Manufacturer Code:NB300232
Catalog # NB300232
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to the C-terminal region of the human SCP3 protein. [UniProt# Q8IZU3] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: anti-SCP3 MGC71888 SCP3 SCP-3 SCP3 chromosome 3 open reading frame 8 SCP3 COR1 synaptonemal complex protein 3; SCP-3; Synaptonemal complex protein 3
Gene Aliases: COR1; RPRGL4; SCP3; SPGF4; SYCP3
UniProt ID: (Human) Q8IZU3
Entrez Gene ID: (Human) 50511
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.