Invitrogen
Manufacturer Code:PA116766
Catalog # APPA6766
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to the C-terminal region of the human SCP3 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: COR1 RNASCP3 Sycp3 Synaptonemal complex protein 3; SCP-3; Synaptonemal complex protein 3; synaptonemal complex protein 3-like
Gene Aliases: COR1; RNASCP3; RPRGL4; SCP3; SPGF4; SYCP3
UniProt ID: (Bovine) Q3T0E2, (Human) Q8IZU3, (Rat) Q63520, (Mouse) P70281
Entrez Gene ID: (Bovine) 615896, (Pig) 100523378, (Human) 50511, (Chicken) 418097, (Rat) 25561, (Mouse) 20962
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.