OriGene
Manufacturer Code:TA507247
Catalog # 50-167-7025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E4 |
Immunogen | Full length human recombit protein of human SCP2 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Acetyl-CoA C-myristoyltransferase; NLTP NSL-TP SCP-2 SCP-CHI SCP-X SCPX; Non-specific lipid-transfer protein; NSL-TP; Propanoyl-CoA C-acyltransferase; SCOX; SCP-2; SCP-2/3-oxoacyl-CoA thiolase; SCP-2/thiolase; SCP-chi; SCP-X; SCPX; Sterol carrier protein 2; Sterol carrier protein X; Straight-chain acyl-CoA oxidase
Gene Aliases: NLTP; NSL-TP; SCP-2; SCP-CHI; SCP-X; SCP2; SCPX
UniProt ID: (Human) P22307
Entrez Gene ID: (Human) 6342
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.