Invitrogen
Manufacturer Code:PA555519
Catalog # PIPA555519
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SCP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetyl-CoA C-myristoyltransferase; Non-specific lipid-transfer protein; nonspecific lipid transfer protein; NSL-TP; Propanoyl-CoA C-acyltransferase; SCOX; SCP-2; SCP-2/3-oxoacyl-CoA thiolase; SCP-2/thiolase; SCP-chi; SCP-X; SCP2 Non-specific lipid-transfer protein Propanoyl-CoA C-acyltransferase SCP-chi SCPX Sterol carrier protein 2 Sterol carrier protein X; SCPX; Sterol carrier protein 2; Sterol carrier protein 2, liver; Sterol carrier protein X; Straight-chain acyl-CoA oxidase
Gene Aliases: AA409774; AA409893; C76618; C79031; NLTP; ns-LTP; NSL-TP; NSLIPTR; SCP-2; SCP-CHI; SCP-X; SCP2; SCPX
UniProt ID: (Human) P22307, (Rat) P11915, (Mouse) P32020
Entrez Gene ID: (Human) 6342, (Rat) 25541, (Mouse) 20280
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.