Invitrogen
Manufacturer Code:PA527354
Catalog # PIPA527354
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 688 and 976 of Human SCP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cancer/testis antigen 8; CT8; CT8 HOM-TES-14 SCP1 SCP-1 cancer/testis antigen 8; SCP-1; Synaptonemal complex protein 1; testicular tissue protein Li 191
Gene Aliases: CT8; HOM-TES-14; SCP-1; SCP1; SYCP1
UniProt ID: (Human) Q15431
Entrez Gene ID: (Human) 6847
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.