Novus Biologicals
Manufacturer Code:NB300228AF350
Catalog # NB30228A350
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoblot (IB) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A C-terminal synthetic peptide made to the human SCP1 protein sequence. [UniProtNo. Q15431] |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
Protein Aliases: Cancer/testis antigen 8; Cancer/testis antigen 8 CT8MGC104417 HOM-TES-14 SCP1SCP-1 synaptonemal complex protein 1; CT8; SCP-1; Synaptonemal complex protein 1; testicular tissue protein Li 191
Gene Aliases: CT8; HOM-TES-14; SCP-1; SCP1; SYCP1
UniProt ID: (Human) Q15431
Entrez Gene ID: (Human) 6847
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.