Invitrogen
Manufacturer Code:PA5117084
Catalog # PIPA5117084
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human SCO2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CEMCOX1 MYP6 Protein SCO2 homolog mitochondrial SCO cytochrome oxidase deficient homolog 2 SCO cytochrome oxidase deficient homolog 2 (yeast) SCO1L Sco2 SCO2 cytochrome c oxidase assembly protein; Protein SCO2 homolog, mitochondrial; SCO cytochrome oxidase deficient homolog 2
Gene Aliases: CEMCOX1; MYP6; SCO1L; SCO2
UniProt ID: (Human) O43819, (Mouse) Q8VCL2
Entrez Gene ID: (Human) 9997, (Mouse) 100126824
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.