Invitrogen
Manufacturer Code:PA555451
Catalog # PIPA555451
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SCNN1D |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Amiloride-sensitive sodium channel subunit delta; Delta-ENaC; Delta-NaCH; ENaCd ENaCdelta SCNED dNaCh amiloride-sensitive sodium channel subunit delta delta-ENaC delta-NaCH epithelial Na(+) channel subunit delta nonvoltage-gated sodium channel 1 subunit delta sodium channel voltage-gated type I delta polypeptide; Epithelial Na(+) channel subunit delta; Nonvoltage-gated sodium channel 1 subunit delta; SCNED; sodium channel, non voltage gated 1 delta subunit; sodium channel, non-voltage-gated 1, delta subunit; sodium channel, nonvoltage-gated 1, delta; sodium channel, voltage-gated, type I, delta polypeptide
Gene Aliases: DNACH; ENaCd; ENaCdelta; SCNED; SCNN1D
UniProt ID: (Human) P51172
Entrez Gene ID: (Human) 6339
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.