Invitrogen
Manufacturer Code:PA577738
Catalog # PIPA577738
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)EKEGQTEYMDY(K) corresponding to amino acid residues 1533-1543 of rat NaV 1.7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Peripheral sodium channel 1; PN1; PN1 Type IX Voltage-Gated Sodium Channel NENA NE Scn9a; Sodium channel protein type 9 subunit alpha; Sodium channel protein type IX subunit alpha; sodium channel, voltage-gated, type 9, alpha polypeptide; Voltage-gated sodium channel subunit alpha Nav1.7
Gene Aliases: Nav1.7; PN1; Scn2a; Scn9a
UniProt ID: (Rat) O08562
Entrez Gene ID: (Rat) 78956
Molecular Function: calcium channel ion channel sodium channel transporter voltage-gated calcium channel voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.