Invitrogen
Manufacturer Code:PA577742
Catalog # PIPA577742
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)KNDKSDPKVRVKDD corresponding to amino acid residues 85-98 of rat NaV beta4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: sodium channel accessory subunit; sodium channel beta 4 subunit; Sodium channel subunit beta-4; sodium channel, type 4, beta subunit; sodium channel, type IV, beta; sodium channel, type IV, beta polypeptide; sodium channel, voltage gated, type IV beta subunit; sodium channel, voltage-gated, type IV, beta subunit; voltage gated sodium channel beta 4 subunit
Gene Aliases: ATFB17; Gm1471; LQT10; Navbeta4; SCN4B
UniProt ID: (Human) Q8IWT1, (Rat) Q7M730, (Mouse) Q7M729
Entrez Gene ID: (Human) 6330, (Rat) 315611, (Mouse) 399548
Molecular Function: cell adhesion molecule ion channel sodium channel transporter voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.