Novus Biologicals
Manufacturer Code:NBP293760100UL
Catalog # NB029382
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of mouse SCN4B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: sodium channel beta-4 subunit sodium channel subunit beta-4 sodium channel voltage-gated type IV beta; Sodium channel subunit beta-4; sodium channel, voltage gated, type IV beta subunit; sodium channel, voltage-gated, type IV, beta subunit
Gene Aliases: ATFB17; LQT10; Navbeta4; SCN4B
UniProt ID: (Human) Q8IWT1
Entrez Gene ID: (Human) 6330
Molecular Function: cell adhesion molecule ion channel sodium channel transporter voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.