Novus Biologicals
Manufacturer Code:NBP259318FR
Catalog # NBP259318FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S396-29 |
Immunogen | Fusion protein amino acids 1-215 (full-length) of rat NavBeta3 |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: HSA243396 KIAA1158 SCNB3 sodium channel subunit beta-3 sodium channel voltage-gated type III beta voltage-gated sodium channel beta-3 subunit; Sodium channel subunit beta-3; sodium channel, voltage gated, type III beta subunit; sodium channel, voltage-gated, type III, beta subunit; voltage-gated sodium channel beta-3 subunit
Gene Aliases: ATFB16; BRGDA7; HSA243396; KIAA1158; SCN3B; SCNB3
UniProt ID: (Human) Q9NY72
Entrez Gene ID: (Human) 55800
Molecular Function: ion channel sodium channel transporter voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.